missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Amyloid beta-peptide (25-35) (human)

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Amyloid β-peptide (25-35) (human) is a fragment of human amyloid β-peptide, functionally required for the neurotrophic and neurotoxic effects associated with Alzheimer's disease.
Specifications
Specifications
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction