missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ANA 12

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TrkB receptor antagonist. Demonstrates a 2-site mode of action, preventing activation by BDNF non-competitively (IC50 values are 45.6 nM and 41.1 μM for the high and low affinity sites respectively). Displays no effect on TrkA and TrkC in an assay of neurite outgrowth. Demonstrates antidepressant properties. Also exhibits anti-seizure effects in an ischemia-induced neonatal seizure model by blocking down-regulation of KCC2.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 407.49 |
| Purity | >99% |
| Molecular Weight (g/mol) | 407.49 |
| Chemical Name or Material | ANA 12 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction