missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ XUBA Analog Ultrasonic Bath

Product Code. p-8014604
Click to view available options
Capacity (Metric):
1.5 L
2.5 L
Unit Size:
Each

Product Code. 11420998

Brand: Grant Instruments™ XUBA1

Only null left
Add to basket
Add to basket

Provides a high standard of reliable and effective ultrasonic technology. Grant Instruments™ XUBA Analog Ultrasonic includes M2 cleaning solution, stainless steel basket and ABS plastic lid included as standard.

  • Excellent entry level ultrasonic baths
  • Temperature range ambient +5°C to 70°C on the XUBA3
  • Fast, effective, efficient, easy and safe cleaning and processing of diverse instruments, components and solutions
  • Supplied with stainless steel basket and ABS plastic lid as standard
  • One bottle of M2 ultrasonic cleaning solution included as standard
  • Control panel easy to operate even when wearing gloves
  • Timer control from 0-15 minutes
TRUSTED_SUSTAINABILITY

Specifications

Timer 0 to 15 min.
Voltage 230 V
Capacity (Metric) 1.5 L
compliance-icons
Product Identifier
  • Grant M2 ultrasonic solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : C9XYLATE, QUATERNARY FATTY AMINE ETHOXYLATE, BENZENESULFONIC ACID
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.