Learn More
Leica Microsystems Analyzer, Rotatable
Product Code. 12887945
Click to view available options
Diameter (Metric):
58 mm
Unit Size:
Each
Product Code. 12887945
Brand: Leica Microsystems 10315306
This item requires a price update prior to being ordered.
Request a Price for this item here.
Improve image visualization with polarization. Leica Microsystems™ Analyzer, Rotatable acts as the second of two filters in the polarization process.
  • Improves image clarity via polarization.
  • Rotatable.
Specifications
Type Analyzer, Rotatable
Diameter (Metric) 58 mm
Quantity 1 Ea.