missing translation for 'onlineSavingsMsg'
Learn More

Anamorelin, MedChemExpress

Product Code. 16471746
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471746 500 mg 500mg
16461136 200 mg 200mg
16458747 5 mg 5mg
16436148 10 mg 10mg
16416536 100 mg 100mg
16412287 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16471746 Supplier Medchem Express Supplier No. HY14734
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Anamorelin (RC-1291) is a potent ghrelin receptor agonist with EC50 value of 0.74 nM in the FLIPR assay.

Anamorelin (RC-1291) is a potent ghrelin receptor agonist with EC50 value of 0.74 nM in the FLIPR assay.

Specifications

Chemical Name or Material Anamorelin
CAS 249921-19-5
Color White
Purity Grade Notes Research
Molecular Formula C31H42N6O3
Quantity 500 mg
Synonym RC-1291 ONO-7643
Solubility Information DMSO : ≥ 100 mg/mL (182.92 mM)
SMILES [H][C@](CC1=CNC2=CC=CC=C21)(NC(C(C)(C)N)=O)C(N3CCC[C@@](C(N(C)N(C)C)=O)(CC4=CC=CC=C4)C3)=O
Molecular Weight (g/mol) 546.7
Formula Weight 546.7
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.