missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Anandamide

Product Code. 18722584
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18722584 5 mg 5mg
18762594 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18722584 Supplier Tocris Bioscience™ Supplier No. 1339/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Endogenous and non-selective CB agonist

Anandamide is an endogenous cannabinoid and TRPV1 receptor agonist (Ki values are 89 and 371 nM at CB1 and CB2 receptors respectively; EC50 values are 18, 31 and 27 nM at GPR55, CB1 and CB2 respectively; pKi = 5.68 for rTRPV1). Also blocks TNF-α-induced NF-κB activation via direct inhibition of IKK. Deuterated analog also available.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Hydroxyethyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
CAS 94421-68-8
Quantity 5 mg
Target Non-selective Cannabinoid Receptor Agonists
Molecular Formula C22H37NO2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.