missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Anandamide
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
Anandamide is an endogenous cannabinoid and TRPV1 receptor agonist (Ki values are 89 and 371 nM at CB1 and CB2 receptors respectively; EC50 values are 18, 31 and 27 nM at GPR55, CB1 and CB2 respectively; pKi = 5.68 for rTRPV1). Also blocks TNF-α-induced NF-κB activation via direct inhibition of IKK. Deuterated analog also available.
Specifications
Specifications
| Chemical Name or Material | N-(2-Hydroxyethyl)-5Z,8Z,11Z,14Z-eicosatetraenamide |
| CAS | 94421-68-8 |
| Quantity | 5 mg |
| Target | Non-selective Cannabinoid Receptor Agonists |
| Molecular Formula | C22H37NO2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction