Learn More
Abeba™ Anatom 2130 Shoes
Product Code. 15637827
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15637827 37 1 pair
15627827 36 1 pair
15647827 38 1 pair
15657827 39 1 pair
15667827 40 1 pair
15677827 41 1 pair
15687827 42 1 pair
15697827 43 1 pair
15607837 44 1 pair
15617837 45 1 pair
15627837 46 1 pair
15637837 47 1 pair
15647837 48 1 pair
15808637 49 1 pair
15818637 50 1 pair
15828637 51 1 pair
15838637 52 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 15637827 Supplier Abeba™ Supplier No. 2130/37
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • Suitable for heel and foot lift
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRC
Color White
Material Microfiber
Size 37
Sole Features Slip-resistant
Sole Material Polyurethane
Style 2130
Toe Material Steel