missing translation for 'onlineSavingsMsg'
Learn More

Ancitabine hydrochloride, MedChemExpress

Product Code. 16482489
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482489 1 g 1g
16411486 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482489 Supplier Medchem Express Supplier No. HYN0093
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ancitabine (hydrochloride) is an important antileukemia drugs.

Ancitabine (hydrochloride) is an important antileukemia drugs.

Specifications

Chemical Name or Material Ancitabine hydrochloride
CAS 10212-25-6
Color White
Purity Grade Notes Research
Molecular Formula C9H12ClN3O4
Quantity 1 g
Synonym Cyclocytidine hydrochloride Cyclo-CMP hydrochloride Cyclo-C
Solubility Information H2O : ≥ 50 mg/mL (191.09 mM) ∣DMSO : 25 mg/mL (95.54 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@@H](O)[C@@](O2)([H])[C@@](N(C2=N3)C=CC3=N)([H])O1.[H]Cl JjbwQ0byOjHpJ4gXWRYObRsaR41DaP==
Molecular Weight (g/mol) 261.66
Formula Weight 261.66
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.