missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Angiogenesis Starter Kit

Product Code. 13443269 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13443269 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13443269 Supplier Gibco™ Supplier No. A1460901
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Angiogenesis Starter Kit

The Gibco™ Angiogenesis Starter Kit contains the reagents and protocols necessary to perform various angiogenesis assays. Designed to address the needs of both the occasional user as well as the most experienced researcher, this combination of high quality products is easy to use and cost effective. Detailed protocols for the tube formation assay and cell migration are available below.

What’s Included:

  • Gibco™ Large Vessel Endothelial Supplement (LVES), 50X, 11 ml, ready to use
  • Gibco™ Medium 200 basal media, 500 ml
  • Gibco™ Human Umbilical Vein Endothelial Cells (HUVEC), 1 vial, 5 x 105 cells
  • Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix, 5 ml

    Large Vessel Endothelial Supplement and Medium 200
    Gibco™ LVES is a 50X, complete, ready-to-use media supplement optimized for use with Gibco™ Medium 200. This blended supplement has been designed to deliver optimal cell growth and to enable superior cell performance for angiogenesis applications such as cell migration and tube formation. LVES contains fetal bovine serum, hydrocortisone, human epidermal growth factor, basic fibroblast growth factor, heparin, and ascorbic acid.

    Human Umbilical Vein Endothelial Cells
    Gibco™ Human Umbilical Vein Endothelial Cells are cryopreserved at the end of the primary culture. Upon thawing, they are guaranteed to be >70% viable and capable of a minimum of 16 population doublings. Cells are derived from tissue obtained from accredited institutions with IRB-approved donor programs. Quality control of these cells includes testing for mycoplasma, sterility, endotoxin, hepatitis B and C, and HIV. Additionally, the cells are positive for von Willebrand factor (vWf), positive for CD31, negative for a-actin, and positive for dil-Ac-LDL uptake.

    Geltrex™ LDEV-Free Matrix
    Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix is a soluble, reduced growth factor, lactate dehydrogenase-elevating virus-free basement membrane extract used in a wide variety of cell culture applications for promotion and maintenance of many cell types, including primary epithelial cells, endothelial cells, and stem cells. It can be diluted for use as a coating matrix for culture vessels or used undiluted as a thick gel preparation to create a more physiologically relevant environment for angiogenesis assays.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Cell Type Endothelial Cells (Umbilical Vein)
    Color Green
    Content And Storage 1 bottle M200500 Medium
    Store at 2–8°C

    1 bottle LVES
    Store at 2–8°C

    1 vial HUCEV
    Store in liquid nitrogen

    1 vial Geltrex LDEV-Free Matrix
    Store at -20 to -80°C
    Detection Method Fluorescence
    For Use With (Application) Angiogenesis
    For Use With (Equipment) Microscope
    Format Cell Culture Plate (Multi-Well)
    Label or Dye Calcein
    Quantity 1 kit
    Type Angiogenesis Starter Kit

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.