missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Angiotensin II 5-valine

Product Code. 16419368
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16419368 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419368 Supplier MedChemExpress Supplier No. HYP0108
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Angiotensin II 5-valine is an agonist of angiotensin receptor.

Specifications

Purity 95.9%
CAS 58-49-1
Content And Storage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 1032.15 g/mol
Product Type Peptides
Quantity 10 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (96.89 mM∣Need ultrasonic)
Synonym Valine angiotensin II∣ 5-L-Valine angiotensin II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.