missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Angiotensin II 5-valine

Product Code. 16479196
Encompass
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479196 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479196 Supplier MedChemExpress Supplier No. HYP0108

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Angiotensin II 5-valine is an agonist of angiotensin receptor.

Specifications

Purity 95.9%
CAS 58-49-1
Content And Storage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 1032.15 g/mol
Product Type Peptides
Quantity 100 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (96.89 mM∣Need ultrasonic)
Synonym Valine angiotensin II∣ 5-L-Valine angiotensin II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.