missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Angiotensin II (human)

Product Code. 15423569
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423569 25 mg 25mg
15413569 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423569 Supplier Thermo Scientific Chemicals Supplier No. J60866.MB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4474-91-31046.197 g/mol

Ang II stimulates angiogenesis and increases microvessel density. In addition, the renin-angiotensin and nitric oxide-generating systems appear to interract in the regulation of cardiovascular function. Angiotensin (Ang) II is a peptide which plays an important role in the regulation of blood pressure and has strong vasoconstrictive effects. It also stimulates the release of aldosterone from the adrenal gland and inhibits adenylate cyclase activity. It also increases the entry of Ca2+ in heart muscle via voltage-sensitive channels and activates myosin light chain kinase (MYLK). It also elicits an increase in voltage-dependent delayed rectifier K+ current in neurons and activates JAK2 in smooth muscle cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ang II stimulates angiogenesis and increases microvessel density. In addition, the renin-angiotensin and nitric oxide-generating systems appear to interract in the regulation of cardiovascular function. Angiotensin (Ang) II is a peptide which plays an important role in the regulation of blood pressure and has strong vasoconstrictive effects. It also stimulates the release of aldosterone from the adrenal gland and inhibits adenylate cyclase activity. It also increases the entry of Ca2+ in heart muscle via voltage-sensitive channels and activates myosin light chain kinase (MYLK). It also elicits an increase in voltage-dependent delayed rectifier K+ current in neurons and activates JAK2 in smooth muscle cells.

Solubility
Soluble in Water (25 mg/ml), and 5% Acetic Acid (1 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. keep away from Strong acids, Strong bases .
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4474-91-3

Specifications

Color White
Content And Storage Store at -20°C
Format Powder
Molecular Weight (g/mol) 1046.197
Product Type Angiotensin II (human)
Quantity 25 mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.