missing translation for 'onlineSavingsMsg'
Learn More

Angiotensin II Trifluoroacetic Acid Salt (Human), TRC

Product Code. 30137967
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137967 1 mg 1mg
30134236 50 mg 50mg
30012591 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137967 Supplier Toronto Research Chemicals Supplier No. TRCA6379651MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C50H71N13O12 . x(C 2HF3O2)
Synonym N-[1-[N-[N-[N-[N-(N2-L-α-Aspartyl-L-arginyl)-L-valyl]-L-tyrosyl]-L-isoleucyl]-L-histidyl]-L-prolyl]-L-phenylalanine, [Ile5]-Ang II, [Ile5]-Angiotensin II, 5-L-Isoleucineangiotensin II, 10: PN: US20130023475 SEQID: 17 unclaimed protein, 10: PN: WO0212471 SEQID: 17 unclaimed sequence, 113: PN: WO2008104000 TABLE: 4 unclaimed sequence, 11: PN: US20030119021 SEQID: 10 unclaimed sequence, 1272: PN: US20050069911 SEQID: 1272 unclaimed sequence, 12: PN: US20090088553 SEQID: 86 unclaimed protein, 12: PN: WO0239997 SEQID: 17 unclaimed sequence, 134: PN: WO02098448 SEQID: 144 unclaimed sequence, 134: PN: WO02098906 SEQID: 144 unclaimed sequence, 13: PN: US20120071544 SEQID: 7 unclaimed sequence, 14: PN: WO2004007538 SEQID: 13 claimed sequence, 14: PN: WO2011020056 SEQID: 95 claimed protein, 154: PN: WO2009126292 TABLE: 4 unclaimed sequence, 17: PN: US6610497 SEQID: 17 unclaimed sequence, 17: PN: US6884771 SEQID: 17 unclaimed sequence, 31: PN: WO0198325 SEQID: 32 claimed protein, 32: PN: US20060014212 SEQID: 32 unclaimed sequence, 32: PN: WO0056345 SEQID: 32 claimed sequence, 7: PN: JP2005291836 SEQID: 6 unclaimed protein, 7: PN: US20040209344 SEQID: 2 unclaimed sequence, 7: PN: WO03072059 SEQID: 8 unclaimed sequence, 80: PN: US6017693 TABLE: 4 claimed sequence, 83: PN: US20050009742 PAGE: 21 claimed sequence, 85: PN: US20080213898 SEQID: 83 claimed sequence, 873: PN: US20040180380 SEQID: 1272 unclaimed sequence, 8: PN: WO2008138350 SEQID: 8 claimed sequence, 8: PN: WO9958140 SEQID: 1 claimed protein, 96: PN: WO2008058016 SEQID: 96 claimed protein, 9: PN: WO2008005531 SEQID: 17 unclaimed sequence, 9: PN: WO2008106043 TABLE: 6 claimed protein, Angiotensin 2, Angiotensin II, Angiotensin II (human), Angiotensin II (mouse), Angiotensin II [Ile5], Asp-Arg-Val-Tyr-Ile-His-Pro-Phe, Asp1-Ile5-angiotensin II, Human angiotensin II, Ileu5-angiotensin II, Isoleucyl5-angiotensin II
IUPAC Name (2S,5S,8S,11S,14S,17S)-2-((1H-imidazol-4-yl)methyl)-17-amino-5-((S)-sec-butyl)-1-((S)-2-(((S)-1-carboxy-2-phenylethyl)carbamoyl)pyrrolidin-1-yl)-14-(3-guanidinopropyl)-8-(4-hydroxybenzyl)-11-isopropyl-1,4,7,10,13,16-hexaoxo-3,6,9,12,15-pentaazanonadecan-19-oic acid2,2,2-trifluoroacetic acid
SMILES O=C(O)[C@H](CC1=CC=CC=C1)NC([C@H]2N(C([C@H](CC3=CNC=N3)NC([C@H]([C@@H](C)CC)NC([C@H](CC4=CC=C(C=C4)O)NC([C@H](C(C)C)NC([C@H](CCCNC(N)=N)NC([C@H](CC(O)=O)N)=O)=O)=O)=O)=O)=O)CCC2)=O.OC(C(F)(F)F)=O.[F,Cl,Br,I]

Specifications

InChI Formula InChI=1S/C50H71N13O12.C2HF3O2/c1-5-28(4)41(47(72)59-36(23-31-25-54-26-56-31)48(73)63-20-10-14-38(63)45(70)60-37(49(74)75)22-29-11-7-6-8-12-29)62-44(69)35(21-30-15-17-32(64)18-16-30)58-46(71)40(27(2)3)61-43(68)34(13-9-19-55-50(52)53)57-42(67)33(51)24-39(65)66;3-2(4,5)1(6)7/h6-8,11-12,15-18,25-28,33-38,40-41,64H,5,9-10,13-14,19-24,51H2,1-4H3,(H,54,56)(H,57,67)(H,58,71)(H,59,72)(H,60,70)(H,61,68)(H,62,69)(H,65,66)(H,74,75)(H4,52,53,55);(H,6,7)/t28-,33-,34-,35-,36-,37-,38-,40-,41-;/m0./s1
Quantity 1 mg
Chemical Name or Material Angiotensin II Trifluoroacetic Acid Salt (Human)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.