Learn More
Buerkle™ Spigot Stopcock™ Angled
Click to view available options
6 to 8mm dia.; Bottles with narrow openings; 1.905cm (0.75in) connection threads
Description
- Tight-closing for complete seal
- Effortless to use
- With connecting cap nut, guaranteed vertical position of the cock
- Pressure-resistant up to 2 bar at 20°C (without discharge bracket and grommet)
- With discharge angle and rotatable spout preventing drips
- With tapered spout for bottles with narrow openings with a diameter from 6 to 8mm
- Media-contacting parts PP, PE and PTFE
Specifications
Material | Polypropylene, Polyethylene, PTFE |
Includes | R 0.75 in. Connection threads, connecting cap nut, discharge angle and rotatable tapered spoμt |