Learn More
Tocris Bioscience™ Ani 9
GREENER_CHOICE
Potent and selective TMEM16A (Anoctamin-1; ANO1) blocker
Brand:  Tocris Bioscience™ 6076/50
Encompass
Product Code. 15757206
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Ani 9 is a potent and selective TMEM16A (Anoctamin-1; ANO1) blocker (IC50 = 77 nM). Displays no effect on ANO2 activity (at 10 μM). Also exhibits no effect on intracellular calcium signaling or CFTR and ENaC channel activity (at 30 μM).
Specifications
Specifications
Ani 9
2-(4-Chloro-2-methylphenoxy)-N'-(2-methoxybenzylidene)acetohydrazide
Soluble to 100mM in DMSO
Off-white
356102-14-2
50 mg
Solid
Bioactive Small Molecule