missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ani 9

Description
Ani 9 is a potent and selective TMEM16A (Anoctamin-1; ANO1) blocker (IC50 = 77 nM). Displays no effect on ANO2 activity (at 10 μM). Also exhibits no effect on intracellular calcium signaling or CFTR and ENaC channel activity (at 30 μM).
Specifications
Specifications
| Chemical Name or Material | Ani 9 |
| CAS | 356102-14-2 |
| Synonym | 2-(4-Chloro-2-methylphenoxy)-N'-(2-methoxybenzylidene)acetohydrazide |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C17H17ClN2O3 |
| Formula Weight | 332.78 |
| Physical Form | Solid |
| Color | Off-white |
| Product Type | Bioactive Small Molecule |
| Show More |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?