missing translation for 'onlineSavingsMsg'
Learn More

Aniline-2-sulfonic Acid, 95%

Product Code. 10514054 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10514054 500 g Glass bottle 500g
10147180 25 g Glass bottle 25g
10246993 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10514054 Supplier Thermo Scientific Acros Supplier No. 400645000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-21-1 | C6H7NO3S | 173.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-21-1
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.19
MDL Number MFCD00007705
InChI Key ZMCHBSMFKQYNKA-UHFFFAOYSA-N
Synonym orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
PubChem CID 6926
ChEBI CHEBI:1015
IUPAC Name 2-aminobenzenesulfonic acid
SMILES C1=CC=C(C(=C1)N)S(=O)(=O)O

Specifications

Melting Point >300°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H2NC6H4SO3H
Quantity 500 g
Beilstein 14, 681
Merck Index 15, 6978
Solubility Information Solubility in water: 3100mg/l (23°C)
Formula Weight 173.19
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Aniline-2-sulfonic acid
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.