missing translation for 'onlineSavingsMsg'
Learn More

Aniline, 99+%

Product Code. 15483087
Change view
Click to view available options
Quantity:
100 g
1000 g
5000 g
Unit Size:
1000g
100g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483087 100 g 100g
11417097 1000 g 1000g
11427097 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15483087 Supplier Thermo Scientific Alfa Aesar Supplier No. A14443.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 62-53-3 | C6H7N | 93.13 g/mol

Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents, iron and iron salts.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-53-3
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00007629
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
Synonym benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
PubChem CID 6115
ChEBI CHEBI:17296
IUPAC Name aniline
SMILES NC1=CC=CC=C1

Specifications

Melting Point -6°C
Density 1.021
Boiling Point 183°C to 184°C
Flash Point 70°C (158°F)
Refractive Index 1.586
Quantity 100 g
UN Number UN1547
Beilstein 605631
Merck Index 14,659
Solubility Information Miscible with water.
Formula Weight 93.13
Percent Purity ≥99%
Chemical Name or Material Aniline
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Aniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: anilin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.