missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Aniline Blue, sodium salt, pure, water soluble, high purity biological stain

Product Code. 10226570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10226570 100 g Glass bottle 100g
10656822 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10226570 Supplier Thermo Scientific Chemicals Supplier No. 229661000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 28983-56-4 | C37H26N3Na2O9S3 | 798.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28983-56-4
Molecular Formula C37H26N3Na2O9S3
Molecular Weight (g/mol) 798.79
MDL Number MFCD00058509
InChI Key TUHAIJABPUJAEY-UHFFFAOYSA-K
Synonym Acid Blue 22, C.I. 42755
PubChem CID 76956083
IUPAC Name disodium;4-[4-[[4-(4-sulfoanilino)phenyl]-[4-(4-sulfonatophenyl)azaniumylidenecyclohexa-2,5-dien-1-ylidene]methyl]anilino]benzenesulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1NC1=CC=C(C=C1)C(C1=CC=C(NC2=CC=CC=C2S([O-])(=O)=O)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1S([O-])(=O)=O

Specifications

Absorption Ratio 0.96 to 1.09
Infrared Spectrum Authentic
Lambda Max 594 to 610nm (in pH 3 water)
Packaging Glass bottle
Color Brown/Purple
Solubility Information Solubility in water: soluble.
Quantity 100 g
Physical Form Crystalline Powder
Formula Weight 799.81
Grade Pure
Chemical Name or Material Aniline Blue, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • Aniline Blue, sodium salt
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.