missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Aniline Blue, sodium salt, pure, water soluble, high purity biological stain

Product Code. 10656822 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10656822 25 g Glass bottle 25g
10226570 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656822 Supplier Thermo Scientific Chemicals Supplier No. 229660250
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28983-56-4 | C37H26N3Na2O9S3 | 798.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28983-56-4
Molecular Formula C37H26N3Na2O9S3
Molecular Weight (g/mol) 798.79
MDL Number MFCD00058509
InChI Key TUHAIJABPUJAEY-UHFFFAOYSA-K
Synonym Acid Blue 22, C.I. 42755
PubChem CID 76956083
IUPAC Name disodium;4-[4-[[4-(4-sulfoanilino)phenyl]-[4-(4-sulfonatophenyl)azaniumylidenecyclohexa-2,5-dien-1-ylidene]methyl]anilino]benzenesulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1NC1=CC=C(C=C1)C(C1=CC=C(NC2=CC=CC=C2S([O-])(=O)=O)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1S([O-])(=O)=O

Specifications

Absorption Ratio 0.96 to 1.09
Infrared Spectrum Authentic
Lambda Max 594 to 610nm (in pH 3 water)
Packaging Glass bottle
Color Brown/Purple
Solubility Information Solubility in water: soluble.
Quantity 25 g
Physical Form Crystalline Powder
Formula Weight 799.81
Grade Pure
Chemical Name or Material Aniline Blue, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • Aniline Blue, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: C.I. Acid Blue 93

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.