missing translation for 'onlineSavingsMsg'
Learn More

Aniline Hydrochloride, TRC

Product Code. 30138021
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138021 500 mg 500mg
30132592 1 g 1g
30007426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138021 Supplier Toronto Research Chemicals Supplier No. TRCA662508500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-04-1
Molecular Formula C6 H7 N . Cl H
Molecular Weight (g/mol) 129.59
Synonym Aniline Hydrochloride, Phenylamine Hydrochloride, 4-Aminobenzoic Acid Imp. C (EP) as Hydrochloride, Sodium Cyclamate Imp. B (EP) as Hydrochloride, Fentanyl Imp. F (EP) as Hydrochloride, Mesalazine Imp. K (EP) as Hydrochloride, Trimethoprim Imp. K (EP) as Hydrochloride
IUPAC Name aniline;hydrochloride
SMILES Cl.Nc1ccccc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H7N.ClH/c7-6-4-2-1-3-5-6;/h1-5H,7H2;1H
Formula Weight 129.0345
Chemical Name or Material Aniline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.