missing translation for 'onlineSavingsMsg'
Learn More

Aniline Hydrochloride, TRC

Product Code. 30132592
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132592 1 g 1g
30138021 500 mg 500mg
30007426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132592 Supplier Toronto Research Chemicals Supplier No. TRCA6625081G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-04-1
Formula Weight 129.0345
InChI Formula InChI=1S/C6H7N.ClH/c7-6-4-2-1-3-5-6;/h1-5H,7H2;1H
IUPAC Name aniline;hydrochloride
Molecular Formula C6 H7 N . Cl H
Molecular Weight (g/mol) 129.59
SMILES Cl.Nc1ccccc1
Synonym Aniline Hydrochloride, Phenylamine Hydrochloride, 4-Aminobenzoic Acid Imp. C (EP) as Hydrochloride, Sodium Cyclamate Imp. B (EP) as Hydrochloride, Fentanyl Imp. F (EP) as Hydrochloride, Mesalazine Imp. K (EP) as Hydrochloride, Trimethoprim Imp. K (EP) as Hydrochloride
Chemical Name or Material Aniline Hydrochloride

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.