missing translation for 'onlineSavingsMsg'
Learn More

Aniline, TRC

Product Code. 30133363
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133363 10 g 10g
30132670 1 g 1g
18051320 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133363 Supplier Toronto Research Chemicals Supplier No. TRCA66247510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-53-3
Molecular Formula C6 H7 N
Molecular Weight (g/mol) 93.13
Synonym Aniline, Aminobenzene, Aminophen, Anyvim, Benzene, amino-, Blue Oil, C.I. 76000, NCI 176889, Phenylamine, Benzenamine
IUPAC Name aniline
SMILES Nc1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2
Formula Weight 93.0578
Percent Purity >95%
Chemical Name or Material Aniline
Grade HPLC
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.