missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Animal-Free Human Dendritic Cell Cytokine Package, PeproTech®

Product Code. 17806687 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17806687 1 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17806687 Supplier Gibco™ Supplier No. AFHDC
  • ONLINE EXCLUSIVE PRICE
    1000.00 €
    860.00 € / Pack of 1
    Save 140.00 €
    14% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Animal-Free Human Dendritic Cell Cytokine Package contains the key components required for the culturing of human dendritic cells.

The Animal-Free Human Dendritic Cell Cytokine Package contains the key components required for the culturing of human dendritic cells.

The package contains the following two vials:
• 100 μg Animal-Free Human IL-4 (Cat. No. F-200-04-100UG)
• 100 μg Animal-Free Human GM-CSF (Cat. No. AF-300-03-100UG)

IL-4 is a pleiotropic cytokine that regulates diverse T and B cell responses including cell proliferation, survival, and gene expression. Produced by mast cells, T cells, and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells.

GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional Assay
Formulation Lyophilized
Product Type Dendritic Cell Cytokine Package
Quantity 1 Pk.
Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.