missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Anios TS Inox Surface Disinfectant

Product Code. 15388934
Change view
Click to view available options
Volume (Metric):
750 mL
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15388934 750 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15388934 Supplier Laboratoires Anios™ Supplier No. 2437241

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Laboratoires Anios™ Anios TS Inox Surface Disinfectant is a double action disinfectant spray for use on stainless steel surfaces.

  • Double action disinfectant cleaner
  • Suitable for use with stainless steel
  • Bactericidal
  • Fungicide
  • Active on HIV-1, Rotavirus, BVDV (model virus HCV) and PRV (HBV model virus)

Specifications

Certifications/Compliance EN 13727, EN 1276, EN 13697, EN 13624, EN 13697
Composition Didecyldimethylammonium chloride (CAS No. 7173-51-5: 1.1mg/g), excipients
For Use With (Application) Cleaning and disinfection of surfaces
Quantity 12/Cs.
Volume (Metric) 750 mL
compliance-icons
Product Identifier
  • anios TS inox premium
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: lactic acid, glycolic acid, N-(2-hydroxyethyl)-N,N-dimethylalkyl-C12-C14-1-aminium chloride, didecy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.