missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniosafe Manuclear HF Soap

Product Code. 15300907
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15300907 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15300907 Supplier Laboratoires Anios™ Supplier No. 1919052

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Laboratoires Anios™ Aniosafe Manuclear HF Soap is a gentle soap recommended for frequent hand hygiene and general body cleansing in hospital environments.

  • Dermatologically tested
  • Enriched with moisturizer
  • Adapted to sensitive skin and for frequent use
  • Preservation of the product demonstrated according to NF EN ISO 11930

Specifications

Product Type Gentle Soap
Color Pink
Certifications/Compliance NF EN ISO 11930
Composition Water, Cocamidopropyl Betaine, Glycerin, Sodium C14-17 Alkyl Sec Sulfonate, PEG-200 Hydrogenated Glyceryl Palmate (and) PEG-7 Glyceryl Cocoate, Benzyl Alcohol, Sodium Benzoate, Sodium Chloride, Lactic acid, dye CI 14720, fragrance
For Use With (Application) General body cleansing in hospital environments
Packaging Bottles with screw pump
Quantity 6 Bottles
Volume (Metric) 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.