Learn More
Laboratoires Anios™ Aniosafe Savon Doux HF
Product Code. 15585290
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Pack of 12
Product Code. Volume (Metric) Unit Size
15585290 1 L Pack of 12
1 options
Product Code. 15585290 Supplier Laboratoires Anios™ Supplier No. 1918229VB
  • ONLINE EXCLUSIVE PRICE
    160.70 €
    141.93 € / Pack of 12
    Save 18.77 €
    11% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Laboratoires Anios™ Aniosafe Savon Doux HF is a gentle soap recommended for frequent hand hygiene and general body cleansing in hospital environments.
  • Dermatologically tested
  • Enriched with moisturizer
  • Preservation of the product demonstrated according to NF EN ISO 11930
Specifications
Product Type Gentle Soap
Certifications/Compliance NF EN ISO 11930
Composition Water, Disodium Lauroamphodiacetate, Sodium Lauryl Sulfate, Hexylene Glycol, Glycerin, Linoleamide DEA, Disodium Ricinoleamido EASulfosuccinate, Benzyl Alcohol, Sodium Benzoate, PEG-200 Hydrogenated Glyceryl Palmate and PEG-7 Glyceryl Cocoate, Parfum (Fragrance), Lactic acid.
For Use With (Application) General body cleansing in hospital environments
Packaging Bottles with screw pump
Quantity 12 Bottles
Volume (Metric) 1 L