missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniosgel 800 Sanitizer

Product Code. 15561823
Change view
Click to view available options
Packaging:
Botte with Screw Pump
Bottles with screw pump
Unit Size:
Pack of 12
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Unit Size
15561823 Botte with Screw Pump Pack of 12
15551823 Bottles with screw pump Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15561823 Supplier Laboratoires Anios™ Supplier No. 2550763GP
  • ONLINE EXCLUSIVE PRICE
    221.60 €
    199.13 € / Pack of 12
    Save 22.47 €
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratoires Anios™ Aniosgel 800 Sanitizer is suitable for hygienic treatment and surgical disinfection of hands by rubbing.

  • High level of observance
  • Tested under dermatological control
  • Adapted to sensitive skin
  • Preserves skin hydration
  • Broad antimicrobial spectrum from 30 sec.
  • Conforms to latest European standards
  • Active against bacteria, mycobacteria, yeasts, molds, viruses

Specifications

Product Type Hand Sanitizer
Form Gel
Certifications/Compliance (Europe) EN 13727, EN 13727, EN14348, EN 13624, EN 14476
Composition Ethanol in the presence of thickening, moisturizing and emollient agents, and water.
For Use With (Application) Hygienic treatment and surgical disinfection of hands by rubbing.
Packaging Botte with Screw Pump
Quantity 12 Bottles
Volume (Metric) 1 L
compliance-icons compliance-icons
Product Identifier
  • aniosgel 800
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403-Store in a well-ventilated place.
  • P501-Dispose of contents/container to as hazardous waste.
Supplemental information
  • MIXTURE LIST-Contain: ethyl alcohol, propan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.