missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniosgel 85 Blue

Product Code. 15441121
Change view
Click to view available options
Volume (Metric):
75 mL
500 mL
Unit Size:
Pack of 12
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15441121 500 mL Pack of 12
15431121 75 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15441121 Supplier Laboratoires Anios™ Supplier No. 1645363

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Laboratoires Anios™ Aniosgel 85 Blue is a hydroalcoholic gel for hygienic treatment and disinfection of hands.

  • Broad antimicrobial spectrum
  • Hygienic treatment in 30 sec.
  • For hygienic treatment and surgical disinfection
  • Active against bacteria, tuberculosis, fungus, viruses

Specifications

Product Type Sterilizer/Disinfectant Hand Gel
Certifications/Compliance (Europe) EN 1040, pr EN 12054, EN 1500, EN 12791, EN 14348, EN 1275, EN 1650, EN 14476+A1
Composition Ethanol in the presence of thickening, moisturizing and emollient agents, and water.
For Use With (Application) Hygienic treatment and surgical disinfection of hands
Packaging Airless bottles
Volume (Metric) 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.