missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniosgel 85 NPC

Product Code. 11988104
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11988104 100 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11988104 Supplier Laboratoires Anios™ Supplier No. 1644505
  • ONLINE EXCLUSIVE PRICE
    108.40 €
    91.63 € / Pack of 20
    Save 16.77 €
    15% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thixotropic hydroalcoholic gel for hygienic treatment and surgical disinfection by hand-rubbing.

Disinfecting gel for hydroalcoholic hand-rubbing:

  • Ethyl alcohol: 755 ml/l
  • Flash point: +23°C
  • Density at +20°C: approx. 0.862
  • pH at +20°C: approx. 5.5 (near to skin pH)
  • Viscosity at +20°C: from 1200 to 2500 mPa.s
  • Storage: from +5°C and +25°C
  • Aerobiocontamination study available on request

Specifications

Product Type Hydroalcoholic Antiseptic Gel
Form Gel
For Use With (Application) For Hygienic Treatment and Antisepsis Surgical Hands
Quantity 20 Pk.
Volume (Metric) 100 mL
compliance-icons compliance-icons
Product Identifier
  • ANIOSGEL 85 NPC
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403-Store in a well-ventilated place.
Supplemental information
  • MIXTURE LIST-Contain: ETHYL ALCOHOL, PROPAN-2-OL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.