missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniosgel 85 NPC

Product Code. 15680105
Change view
Click to view available options
Volume (Metric):
500 mL
1 L
Packaging:
Airless bottles
Pump Bottles
Unit Size:
Pack of 12
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Packaging Unit Size
15680105 500 mL Pump Bottles Pack of 12
15670105 1 L Airless bottles Pack of 12
15600115 1 L Pump Bottles Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15680105 Supplier Laboratoires Anios™ Supplier No. 1644748
  • ONLINE EXCLUSIVE PRICE
    143.30 €
    120.15 € / Pack of 12
    Save 23.15 €
    16% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratoires Anios™ Aniosgel 85 NPC contains no allergenic substances, fragrance, or coloring, and is suitable for hygienic treatment and surgical disinfection of hands

  • No allergenic substances, fragrance, or coloring
  • Broad antimicrobial spectrum
  • Hygienic treatment in 30 sec.
  • For hygienic treatment and surgical disinfection
  • Active against bacteria, mycobacteria, yeasts, molds, viruses

Specifications

Product Type Hand Sterilizer/Disinfectant
Form Gel
Certifications/Compliance (Europe) EN 13727, EN 13727, EN 14348, EN 13624, EN 14476, EN 14476, EN 1500, EN 12791
Composition Ethanol in the presence of thickening, moisturizing and emollient agents, and water.
For Use With (Application) Hygienic treatment and surgical disinfection of hands
Packaging Pump Bottles
Quantity 12 Bottles
Volume (Metric) 500 mL
compliance-icons compliance-icons
Product Identifier
  • ANIOSGEL 85 NPC
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403-Store in a well-ventilated place.
Supplemental information
  • MIXTURE LIST-Contain: ETHYL ALCOHOL, PROPAN-2-OL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.