missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniospray Quick Spray Surface Disinfectant

Product Code. 15461884
Change view
Click to view available options
Volume (Metric):
1 L
5 L
Quantity:
12/Cs.
4/Cs.
Unit Size:
Pack of 12
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Quantity Unit Size
15461884 5 L 4/Cs. Pack of 4
15451884 1 L 12/Cs. Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461884 Supplier Laboratoires Anios™ Supplier No. 2084034
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratoires Anios™ Aniospray Quick Spray Surface Disinfectant is an ethanol-based, broad antimicrobial spectrum spray for rapid disinfection of surfaces and non-immersible, non-invasive equipment.

  • Ethanol-based composition
  • Broad activity spectrum
  • Leaves no trace when drying
  • Pleasant fragrance
  • Aldehyde free, without coloring
  • Rapid action disinfection of surfaces and previously cleaned, non-immersible medical devices resistant to alcohol (stethoscopes, pressure sensors, blood sugar testers, etc.)
  • Active against bacteria, mycobacteria, yeasts, molds, viruses

Specifications

Product Type Rapid Disinfectant
Certifications/Compliance Bacteria: EN 13727, EN 1276, EN 13697, MRB according to EN 13697Mycobacteria: EN 14348, EN 14563Yeasts/Molds: EN 13624, EN 1650, EN 13697, EN 14562 : Tricophyton mentagrophytesViruses: Adenovirus, HIV-1, PRV (surrogate of HBV), BVDV (surrogate of HCV), Vaccinia virus Herpes-virus, Coronavirus, Rotavirus, Infl uenza virus A (H N ), Feline Calicivirus, Norovirus MNV according to EN 14476
Composition Hydroalcoholic solution (ethanol 55%), quaternary ammonium propionate,fragrance.
For Use With (Application) Rapid disinfection of non-immersible medical devices resistant to alcohol.
Quantity 4/Cs.
Volume (Metric) 5 L
compliance-icons compliance-icons
Product Identifier
  • ANIOSPRAY QUICK
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ETHYL ALCOHOL, PROPAN-2-OL, DIDECYLDIMETHYLAMMONIUM CHLORIDE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.