missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniospray Surf 29 Disinfectant

Product Code. 15348944
Change view
Click to view available options
Quantity:
12/Cs.
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15348944 12/Cs. Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15348944 Supplier Laboratoires Anios™ Supplier No. 2421073
  • ONLINE EXCLUSIVE PRICE
    187.40 €
    157.35 € / Pack of 12
    Save 30.05 €
    16% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A ready-to-use disinfectant spray for rapid disinfection of surfaces, medical equipment and previously cleaned non-immersible and non-invasive medical devices (e.g. stethoscopes, pressure sensors).

  • Aldehyde-free
  • Bactericidal and fungicidal according to the standard NT F 72-281; active against viruses
  • Composition of ethanol and didecyldimethylammonium chloride, chlorhexidine digluconate
  • Use undiluted on surfaces previously cleaned
  • Does not leave any trace or greasy film

Specifications

Composition Ethanol and didecyldimethylammonium chloride, chlorhexidine digluconate
Container Type Bottle
Description Disinfectant spray
For Use With (Application) Disinfection of surfaces and medical equipment
Includes 1 Trigger Sprayer
Quantity 12/Cs.
Volume (Metric) 1 L
compliance-icons compliance-icons
Product Identifier
  • ANIOSPRAY SURF 29
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: ETHYL ALCOHOL, D-GLUCONIC ACID, COMPOUNDWITH N, N''-BIS(4-CHLOROPHENYL)-3, 12-DIIMINO-2, 4, 11, 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.