missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniospray Surf 29

Product Code. 15338944
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15338944 5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15338944 Supplier Laboratoires Anios™ Supplier No. 2421034
  • ONLINE EXCLUSIVE PRICE
    224.90 €
    191.09 € / Pack of 4
    Save 33.81 €
    15% Off
View all promotions
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Disinfection of surfaces, medical equipment and non-submersible and non-invasive medical devices, previously cleaned (stethoscopes, pressure sensors, blood sugar testers, etc.).

Disinfectant spray

  • Ready to use product
  • Clear and colourless solution
  • Density at +20°C : ≈ 0.950
  • pH at +20°C : ≈ 6.0
  • Percentage of ethanol : 41% v/v
  • Flash point : +26.5°C
  • Not corrosive and not oxydising
  • 1 pressure sprayer provides 1 ml product

Specifications

Form Liquid
Composition Ethanol, didecyldimethylammonium chloride, chlorhexidine digluconate, excipients.
Container Type Can
Quantity 4/Cs.
Test Time 5 minutes
Volume (Metric) 5 L
compliance-icons compliance-icons
Product Identifier
  • ANIOSPRAY SURF 29
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: ETHYL ALCOHOL, D-GLUCONIC ACID, COMPOUNDWITH N, N''-BIS(4-CHLOROPHENYL)-3, 12-DIIMINO-2, 4, 11, 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.