missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniosurf ND Premium

Product Code. 15348954
Change view
Click to view available options
Volume (Metric):
1 L
5 L
Quantity:
12/Cs.
4/Cs.
Unit Size:
Pack of 12
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Quantity unitSize
15348954 5 L 4/Cs. Pack of 4
15398934 1 L 12/Cs. Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15348954 Supplier Laboratoires Anios™ Supplier No. 2450036

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Laboratoires Anios™ Aniosurf ND Premium has broad antimicrobial efficiency and is suitable for disinfecting floors, walls, and medical equipment.

  • Cleaning and disinfection of floors, walls and medical equipment
  • Broad antimicrobial efficiency
  • Non-corrosive formula, compatible with washable surfaces
  • Does not leave film or sticky residue
  • Usable in warm and lukewarm water
  • Active against bacteria, mycobacteria, yeasts, molds, viruses

Specifications

Product Type Disinfectant Concentrate
pH 7 ±0.5
Scent Fresh scent
Certifications/Compliance Bacteria: EN 1040, EN 13727Active on BMR and BHR according to EN 13727Active on K. pneumoniae OXA 48, EN 1369Mycobacteria: M. terrae : EN 14348M. terrae : EN 13697Yeasts: EN 1275, EN 13624, EN 13697Molds: EN 13624, EN 13697Viruses: According to EN 14476: PRV model HBV virus), BVDV (HCV model virus), Vacciniavirus, Herpes virus, According to EN 14476: Rotavirus
Composition Didecyldimethylammonium chloride (CAS No. 7173-51-5: 82mg /g), Chlorhexidine digluconate (CAS RN 18472-51-0: 5.2mg /g), excipients
Content And Storage 5°C to 35°C
Dilution Dilute at 0.25%
For Use With (Application) Disinfection of floors, walls and medical equipment
Quantity 4/Cs.
Volume (Metric) 5 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.