missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniosurf ND Premium

Product Code. p-7146818
Click to view available options
Volume (Metric):
1 L
5 L
Quantity:
12/Cs.
4/Cs.
Unit Size:
Pack of 12
Pack of 4
This item is not returnable. View return policy

Product Code. 15348954

Brand: Laboratoires Anios™ 2450036

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Laboratoires Anios™ Aniosurf ND Premium has broad antimicrobial efficiency and is suitable for disinfecting floors, walls, and medical equipment.

  • Cleaning and disinfection of floors, walls and medical equipment
  • Broad antimicrobial efficiency
  • Non-corrosive formula, compatible with washable surfaces
  • Does not leave film or sticky residue
  • Usable in warm and lukewarm water
  • Active against bacteria, mycobacteria, yeasts, molds, viruses

Specifications

Product Type Disinfectant Concentrate
pH 7 ±0.5
Scent Fresh scent
Certifications/Compliance Bacteria: EN 1040, EN 13727Active on BMR and BHR according to EN 13727Active on K. pneumoniae OXA 48, EN 1369Mycobacteria: M. terrae : EN 14348M. terrae : EN 13697Yeasts: EN 1275, EN 13624, EN 13697Molds: EN 13624, EN 13697Viruses: According to EN 14476: PRV model HBV virus), BVDV (HCV model virus), Vacciniavirus, Herpes virus, According to EN 14476: Rotavirus
Composition Didecyldimethylammonium chloride (CAS No. 7173-51-5: 82mg /g), Chlorhexidine digluconate (CAS RN 18472-51-0: 5.2mg /g), excipients
Content And Storage 5°C to 35°C
Dilution Dilute at 0.25%
For Use With (Application) Disinfection of floors, walls and medical equipment
Quantity 4/Cs.
Volume (Metric) 5 L
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : didecyldimethylammonium chloride, propan-2-ol, N,N"-bis(4-chlorophenyl)-3,12-diimino-2,3,11,13-tetra
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.