missing translation for 'onlineSavingsMsg'
Learn More

Anisaldehyde Solution, Fisher Chemical™
BIOPHARMA

Product Code. 12900364 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12900364 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12900364 Supplier Fisher Chemical Supplier No. RP 022 100
  • ONLINE EXCLUSIVE PRICE
    114.64 €
    103.94 € / 100mL
    Save 10.70 €
    9% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

CAS 123-11-5
Molecular Formula C8H8O2
Quantity 100 mL
Synonym p-anisaldehyde, anisaldehyde, anisic aldehyde, p-methoxybenzaldehyde, 4-anisaldehyde, aubepine, benzaldehyde, 4-methoxy, p-anisic aldehyde, p-formylanisole, crategine
InChI Key ZRSNZINYAWTAHE-UHFFFAOYSA-N
Molecular Weight (g/mol) 136.15
PubChem CID 31244
ChEBI CHEBI:28235
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Anisaldehyde solution. European Pharmacopoeia Ref: 1007301. (RP 022)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands, forarms and face thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P311-Call a POISON CENTER/doctor/ .
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: methanol, acetic acid, sulphuric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.