missing translation for 'onlineSavingsMsg'
Learn More

Anisil, 98+%

Product Code. 11473264
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473264 100 g 100g
11453264 5 g 5g
11463264 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473264 Supplier Thermo Scientific Alfa Aesar Supplier No. A10594.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1226-42-2 | C16H14O4 | 270.284 g/mol

Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1226-42-2
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.284
MDL Number MFCD00008405
InChI Key YNANGXWUZWWFKX-UHFFFAOYSA-N
Synonym 4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione
PubChem CID 71043
IUPAC Name 1,2-bis(4-methoxyphenyl)ethane-1,2-dione
SMILES COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC

Tekniske data

Melting Point 132°C to 134°C
Quantity 100 g
Beilstein 531345
Solubility Information Insoluble in water.
Formula Weight 270.29
Percent Purity ≥98%
Chemical Name or Material Anisil

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.