missing translation for 'onlineSavingsMsg'
Learn More

BD Annexin V-FITC

Product Code. 15878688
Change view
Click to view available options
Quantity:
100 Tests
200 Tests
25 Tests
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15878688 100 Tests Each
10186463 200 Tests Each
15850609 25 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15878688 Supplier BD Supplier No. 556420
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Serves as a sensitive probe for flow cytometric analysis of cells that are undergoing apoptosis

Identifies apoptosis at an earlier stage than assays based on nuclear changes such as DNA fragmentation.

  • Format: FITC

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 Tests
Sample Volume 5 μL
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Conjugate FITC

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.