missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Ansamitocin P-3

Product Code. 16419337
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16419337 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419337 Supplier MedChemExpress Supplier No. HY15739
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ansamitocin P-3 (Antibiotic C 15003P3) is a microtubule inhibitor. Ansamitocin P-3 is a macrocyclic antitumor antibiotic.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 635.14 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 66584-72-3
Product Type Natural Products
Purity 98.0%
Solubility DMSO : ≥ 100 mg/mL (157.45 mM)
Synonym Antibiotic C 15003P3∣ Maytansinol isobutyrate
Target Microtubule/Tubulin∣ADC Cytotoxin∣Bacterial∣Antibiotic
Quantity 5 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.