missing translation for 'onlineSavingsMsg'
Learn More

Ansell TouchNTuff™ DermaShield™ 73-701 Neoprene Cleanroom Gloves
BIOPHARMA

Product Code. 12465090 Shop All Ansell Products
Change view
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
Unit Size:
200 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12465090 9 200 pairs
11706713 6 200 pairs
11716713 6.5 200 pairs
11726713 7 200 pairs
11736713 7.5 200 pairs
11746713 8 200 pairs
11756713 8.5 200 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12465090 Supplier Ansell Supplier No. 73701090
  • ONLINE EXCLUSIVE PRICE
    829.00 €
    515.46 € / 200 pairs
    Save 313.54 €
    37% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Sterile neoprene (polychloroprene) cleanroom gloves are free of proteins and accelerators. Suitable for use in aseptic and Class 100/ISO 5 cleanroom environments.

  • Sterile disposable gloves feature a unique neoprene formulation for greater durability and puncture resistance.
  • Offers high levels of chemical splash resistance protecting from various hazards.
  • Beaded cuff designed with SUREFIT™ technology prevents cuffs from rolling down.
  • Latex-free and chemical accelerator-free eliminates Type I and Type IV allergy risks.
  • Recommended for transferring liquids and solids, sample taking, blending, compounding, and aseptic filling/labeling.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance 21 CFR 820 Quality System Regulations, ASTM D3577, ASTM D7160, ASTM D6978-05, ISO 10282, ISO 11193, ISO 13485, ISO 14001
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009, EN 421:2010, EN 556-1:2001, EN ISO 374-1:2016, EN ISO 374-5:2016
Chemotherapeutic Rated Yes
Chlorinated Yes
Cleanroom Class Class 100 (ISO 5)
Accelerator Free Yes
AQL 0.65
Color Green
Disposable Single-use
Exterior Finish No Texture
Finger Thickness (English) 8.3 mil
Finger Thickness (Metric) 0.19 mm
Hand Specific
Latex Free Yes
Material Polychloroprene
Palm Thickness (English) 7 mil
Palm Thickness (Metric) 0.18 mm
Powdered Powder Free
Product Type Cleanroom Gloves
Size 9
Sterility Sterile
Thickness (English) 8.3 mil
Thickness (Metric) 0.19 mm
Grip Design Textured Fingers
Manufacturing/QMS Audit Standards ISO 9001:2015
Model 73-701
Series TouchNTuff 73-701
Tensile Strength 17 MPa
Length (English) 11.6 in.
Length (Metric) 29.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.