missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-501 Aramid Fiber Stretch Armor Gloves, Nitrile Coated

Product Code. 12690692
Change view
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12690692 6 12 pairs
12600702 7 12 pairs
12610702 8 12 pairs
12620702 9 12 pairs
12670692 10 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12690692 Supplier Ansell Supplier No. 11501060
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ergonomic design reduces hand fatigue and provides a fit that mimics bare hands. Nitrile foam coating enhances tactile sensitivity and grip capabilities in dry or light oil conditions.

  • Intercept Technology™ raises the protection & dexterity standards for cut-resistant gloves
  • Aramid fiber provides an additional level of protection
  • Ergonomic design provides bare-hand closeness and reduces hand fatigue
  • Nitrile foam coating enhances grip capabilities and tactile sensitivity
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A5
Hand Specific
Latex Free Yes
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size 6
For Use With (Application) Automotive Repairs, Machinery/Equipment Operation, Metal Fabrication, and Chemical Handling
Glove Cuff Style Knit
Series 11-501
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.