missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-900 Nitrile Coated Oil Repellent Gloves

Product Code. 12670762
Change view
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12670762 6 12 pairs
12680762 7 12 pairs
12690762 8 12 pairs
12600772 9 12 pairs
12660762 10 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12670762 Supplier Ansell Supplier No. 11900060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Industrial work gloves designed for light mechanical protection, particularly in moderately oily manufacturing or maintenance environments.

  • Nitrile coating provides light mechanical protection and dexterity
  • Designed for use in moderately oily manufacturing and maintenance environments
  • Combines protection with enhanced grip and abrasion resistance
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Chlorinated No
Construction Knit
Cuff Material Nitrile
Coating Material Nitrile
Color White, Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 6
For Use With (Application) Assembly, Equipment Operation, and Metal Fabrication
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Lining Unlined
Series 11-900
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.