missing translation for 'onlineSavingsMsg'
Learn More

Antazoline hydrochloride, MedChemExpress

Product Code. 16438586
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438586 Supplier Medchem Express Supplier No. HYB1067
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Antazoline hydrochloride is a 1st generation antihistamine with also anticholinergic properties used to relieve nasal congestion and in eye drops.

Antazoline hydrochloride is a 1st generation antihistamine with also anticholinergic properties used to relieve nasal congestion and in eye drops.

Specifications

Chemical Name or Material Antazoline hydrochloride
CAS 2508-72-7
Color White
Purity Grade Notes Research
Molecular Formula C17H20ClN3
Quantity 100 mg
Synonym Phenazoline hydrochloride
Solubility Information DMSO : ≥ 53 mg/mL (175.61 mM)
SMILES [H]Cl.C1(CN(C2=CC=CC=C2)CC3=CC=CC=C3)=NCCN1
Molecular Weight (g/mol) 301.81
Formula Weight 301.81
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.