Learn More
anti-CD3 zeta, Clone: H146-968, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand:  Novus Biologicals NBP1-44683
Encompass
Product Code. 15798415
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
CD3 zeta Monoclonal specifically detects CD3 zeta in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Flow (Intracellular), CyTOF-ready.