missing translation for 'onlineSavingsMsg'
Learn More

Mouse B Lymphocyte Enrichment Set - DM, BD IMag-DM, Biotin, BD

Product Code. 15889048
Change view
Click to view available options
Conjugate:
BD IMag-DM, Biotin
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Conjugate Unit Size
15889048 BD IMag-DM, Biotin Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15889048 Supplier BD Biosciences Supplier No. 557792
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cell Separation

TRUSTED_SUSTAINABILITY

Specifications

Classification Monoclonal
Conjugate BD IMag-DM, Biotin
Regulatory Status RUO
Primary or Secondary Primary
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.