missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Anti-bacterial, Anti-odor Insoles

Product Code. 17495033
Change view
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17495033 45 1 pair
17425033 38 1 pair
17435033 39 1 pair
17445033 40 1 pair
17455033 41 1 pair
17465033 42 1 pair
17475033 43 1 pair
17485033 44 1 pair
17405043 46 1 pair
17415043 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 17495033 Supplier Honeywell™ Supplier No. 600031045/7
  • ONLINE EXCLUSIVE PRICE
    13.42 €
    7.07 € / 1 pairs
    Save 6.35 €
    47% Off
View all promotions
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Double-action, perforated blue insoles prevent bacteria and odor.

  • Dual-action: prevents bacteria and odor
  • Perforated for breathability
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Design Perforated
For Use With (Application) Preventing Bacteria and Odor
Size 45
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.