missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD229/SLAMF3 Mouse anti-Human, Clone: 249936, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB1898-100
This item is not returnable.
View return policy
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| CD229/SLAMF3/Lymphocyte Antigen 9 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, Ly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4063 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| 249936 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9HBG7 | |
| LY9 | |
| Mouse myeloma cell line NS0-derived recombinant human CD229/SLAMF3 Lys48-Lys454 Accession # Q9HBG7 | |
| 100 μg | |
| Primary | |
| Detects human CD229/SLAMF3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD229 is observed. | |
| Human | |
| Supernatant |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction