missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPB41 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539787
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Elliptocytosis is a hematologic disorder characterized by elliptically shaped erythrocytes and a variable degree of hemolytic anemia. Inherited as an autosomal domit, elliptocytosis results from mutation in any one of several genes encoding proteins of the red cell membrane skeleton. The form discussed here is the one found in the 1950s to be linked to Rh blood group and more recently shown to be caused by a defect in protein 4.1. ′Rh-unlinked′ forms of elliptocytosis are caused by mutation in the alpha-spectrin gene, the beta-spectrin gene, or the band 3 gene.Specifications
EPB41 | |
Polyclonal | |
Unconjugated | |
EPB41 | |
E41P, EPB4.1, band 4.1, P4.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2035, 269587 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P11171, P48193 | |
EPB41 | |
A synthetic non-phosphopeptide derived from human EPB41 around the phosphorylation site of Tyr660/418 (N-I-YP-I-R) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |